Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Lavendustin B | 125697-91-8 | MFCD00133446 | 98.0% | 365.38 | C21H19NO5 | 5 MG
SDP

Catalog No. 5000264401
Encompass_Preferred

Lavendustin B is a small-molecule research reagent that inhibits HIV-1 integrase interaction with LEDGF/p75, functions as an ATP-competitive GLUT1 inhibitor, and exhibits weak tyrosine kinase inhibition. Supplied as a solid for laboratory research use, it is characterized by its CAS number 125697-91-8 and defined molecular properties.

  • High purity (98.0%).
  • Molecular weight 365.38 g/mol.
  • Chemical formula C21H19NO5.
  • Available in small milligram quantities suitable for screening and assays.
  • Stable as powder when stored at -20°C for long-term storage.
  • Suitable for biochemical and cell-based assay applications.

Catalog No. 50-002-64401 Supplier Medchemexpress LLC Supplier No. HY1089355MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Lavendustin B is a small-molecule research reagent that inhibits HIV-1 integrase interaction with LEDGF/p75, functions as an ATP-competitive GLUT1 inhibitor, and exhibits weak tyrosine kinase inhibition. Supplied as a solid for laboratory research use, it is characterized by its CAS number 125697-91-8 and defined molecular properties.

  • High purity (98.0%).
  • Molecular weight 365.38 g/mol.
  • Chemical formula C21H19NO5.
  • Available in small milligram quantities suitable for screening and assays.
  • Stable as powder when stored at -20°C for long-term storage.
  • Suitable for biochemical and cell-based assay applications.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.