Learn More
Medchemexpress LLC Lawsone | 83-72-7 | 25 G
SDP
Catalog No. 5000422021
Encompass_Preferred
Lawsone is an orally active naphthoquinone dye isolated from the leaves of *Lawsonia inermis*. It can induce apoptosis and possesses antibacterial, antitumor, and antioxidant activities, making it suitable for anti-tumor drug research.
  • Induces apoptosis
  • Possesses antibacterial activity
  • Exhibits antitumor activity
  • Has antioxidant properties
  • Inhibits growth of *Escherichia coli* in a dose-dependent manner
  • Inhibits cell proliferation in human DLD-1 cells
  • Has an anti-proliferation effect in SKOV-3 ovarian cancer cells
Catalog No. 50-004-22021 Supplier Medchemexpress LLC Supplier No. 25GHYN2493
Lawsone is an orally active naphthoquinone dye isolated from the leaves of *Lawsonia inermis*. It can induce apoptosis and possesses antibacterial, antitumor, and antioxidant activities, making it suitable for anti-tumor drug research.
  • Induces apoptosis
  • Possesses antibacterial activity
  • Exhibits antitumor activity
  • Has antioxidant properties
  • Inhibits growth of *Escherichia coli* in a dose-dependent manner
  • Inhibits cell proliferation in human DLD-1 cells
  • Has an anti-proliferation effect in SKOV-3 ovarian cancer cells