Learn More
Medchemexpress LLC Lawsone | 83-72-7 | 50 G
SDP
Supplier:  Medchemexpress LLC HYN249350G
Encompass_Preferred
Lawsone is an orally active naphthoquinone dye isolated from the leaves of *Lawsonia inermis*. It is utilized in anti-tumor drug research and exhibits antibacterial, antitumor, and antioxidant activities, as well as inducing apoptosis.
  • Orally active naphthoquinone dye
  • Isolated from the leaves of *Lawsonia inermis*
  • Induces apoptosis
  • Exhibits antibacterial activity
  • Exhibits antitumor activity
  • Exhibits antioxidant activities
  • Used in anti-tumor drug research
  • Solid appearance
  • Light yellow to yellow color
Catalog No. 50-004-20903