Learn More
Medchemexpress LLC Lawsone methyl ether (2-methoxy-1,4-naphthoquinone) | 2348-82-5 | MFCD00019539 | 1 ML
SDP
Catalog No. 5000418076
Encompass_Preferred
Lawsone methyl ether (2-Methoxy-1,4-naphthoquinone) is a natural compound isolated from Impatiens balsamina L. and Swertia calycina, known for its potent antifungal and antibacterial activities. This product is provided as a solution in DMSO.
  • High purity of 99.16%
  • Provided as a convenient solution
  • Concentration of 10 mM
  • Volume of 1 mL
Catalog No. 50-004-18076 Supplier Medchemexpress LLC Supplier No. HYN711610MM/1ML
Lawsone methyl ether (2-Methoxy-1,4-naphthoquinone) is a natural compound isolated from Impatiens balsamina L. and Swertia calycina, known for its potent antifungal and antibacterial activities. This product is provided as a solution in DMSO.
  • High purity of 99.16%
  • Provided as a convenient solution
  • Concentration of 10 mM
  • Volume of 1 mL