Learn More
Aviva Systems Biology Corp LAX1 Antibody - middle region (ARP47347_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP47347_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LAX1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human LAX1. Alternate names: FLJ20340, LAXApplicationWB. Species reactivity: Cow, Human, Rat
Catalog No. 50-014-59518