Learn More
Aviva Systems Biology Corp LAX1 Antibody - C-terminal region (ARP59140_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP59140_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LAX1. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the C terminal region of human LAX1. Alternate names: FLJ20340, LAXApplicationWB. Species reactivity: Dog, Human, Mouse, Pig, Rat
Catalog No. 50-286-591