Learn More
Cayman Chemical LayIlIn Isoform 2 EXrAClul
SDP
Catalog No. 502919366
Encompass_Preferred
Source: Recombinant C-terminal human IgG1 Fc-tagged layilin isoform 2 extracellular domain expressed in HEK293 cells • Amino acids: 22-220 • MW: 49.5 kDa
Catalog No. 50-291-9366 Supplier Cayman Chemical Supplier No. 37021100 UG
Source: Recombinant C-terminal human IgG1 Fc-tagged layilin isoform 2 extracellular domain expressed in HEK293 cells • Amino acids: 22-220 • MW: 49.5 kDa