Learn More
Aviva Systems Biology Corp LAYN Antibody - N-terminal region (ARP53385_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP53385_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against LAYN. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human LAYN. Alternate names: FLJ30977, FLJ31092ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Rabbit, Rat
Catalog No. 50-014-53714