Learn More
Medchemexpress LLC HY-109061 10mg Medchemexpress, Lazertinib CAS:1903008-80-9 Purity:>98%
SDP
Catalog No. 502028791
Encompass_Preferred
Medchemexpress, HY-109061 10mg Lazertinib CAS:1903008-80-9 Lazertinib (YH25448) is a potent, highly mutant-selective, blood-brain barrier permeable, orally available and irreversible third-generation EGFR tyrosine kinase inhibitor, and can be used in the research of non-small cell lung cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-202-8791 Supplier Medchemexpress LLC Supplier No. HY10906110MG
Medchemexpress, HY-109061 10mg Lazertinib CAS:1903008-80-9 Lazertinib (YH25448) is a potent, highly mutant-selective, blood-brain barrier permeable, orally available and irreversible third-generation EGFR tyrosine kinase inhibitor, and can be used in the research of non-small cell lung cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.