Learn More
Selleck Chemical LLC Lazertinib 5mg 1903008-80-9 YH25448,GNS-1480
SDP
Supplier:  Selleck Chemical LLC S87245MG
Encompass_Preferred
Lazertinib (YH25448,GNS-1480) is a potent, highly mutant-selective and irreversible EGFR-TKI with IC50 values of 1.7 nM, 2 nM, 5 nM, 20.6 nM and 76 nM for Del19/T790M, L858R/T790M, Del19, L85R and Wild type EGFR respectively, showing much higher IC50 values aganist ErbB2 and ErbB4. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-194-8203