Learn More
Medchemexpress LLC Lazertinib (mesylate) | 2247995-37-3 | 99.85% | 650.75 | 1 ML
SDP
Catalog No. 5000344626
Encompass_Preferred
Lazertinib mesylate is a potent, selective, CNS-penetrant, orally available, and irreversible EGFR tyrosine kinase inhibitor. It shows high selectivity for activating (EGFRm) and T790M resistance mutations, inhibiting phosphorylation of EGFR, AKT, and ERK. This leads to apoptosis and suppression of tumor growth, making it suitable for the study of non-small cell lung cancer.
  • Potent, selective, CNS-penetrant, orally available, and irreversible EGFR tyrosine kinase inhibitor.
  • Shows high selectivity for activating (EGFRm) and T790M resistance mutations.
  • Inhibits phosphorylation of EGFR, AKT, and ERK.
  • Leads to apoptosis and suppression of tumor growth.
  • Suitable for the study of non-small cell lung cancer.
Catalog No. 50-003-44626 Supplier Medchemexpress LLC Supplier No. HY109061B10MM/1ML
Lazertinib mesylate is a potent, selective, CNS-penetrant, orally available, and irreversible EGFR tyrosine kinase inhibitor. It shows high selectivity for activating (EGFRm) and T790M resistance mutations, inhibiting phosphorylation of EGFR, AKT, and ERK. This leads to apoptosis and suppression of tumor growth, making it suitable for the study of non-small cell lung cancer.
  • Potent, selective, CNS-penetrant, orally available, and irreversible EGFR tyrosine kinase inhibitor.
  • Shows high selectivity for activating (EGFRm) and T790M resistance mutations.
  • Inhibits phosphorylation of EGFR, AKT, and ERK.
  • Leads to apoptosis and suppression of tumor growth.
  • Suitable for the study of non-small cell lung cancer.