Learn More
Cayman Chemical LAzIdohomoalnInhydrochlo 100mg
SDP
Catalog No. 502952122
Encompass_Preferred
A clickable form of L-homoalanine; has been used for semiquantitative analysis of newly synthesized proteins in A549 epithelial cancer cells and neural progenitor cells derived from patients with fragile X syndrome
Catalog No. 50-295-2122 Supplier Cayman Chemical Supplier No. 11786100 MG
A clickable form of L-homoalanine; has been used for semiquantitative analysis of newly synthesized proteins in A549 epithelial cancer cells and neural progenitor cells derived from patients with fragile X syndrome