Learn More
Cayman Chemical L-Azidohomoalanine (hydrochloride), Purity: ≥95%, CAS: 942518-29-8, 250 mg
SDP
Catalog No. 502951959
Encompass_Preferred
A clickable form of L-homoalanine; has been used for semiquantitative analysis of newly synthesized proteins in A549 epithelial cancer cells and neural progenitor cells derived from patients with fragile X syndrome
Catalog No. 50-295-1959 Supplier Cayman Chemical Supplier No. 11786250 MG
A clickable form of L-homoalanine; has been used for semiquantitative analysis of newly synthesized proteins in A549 epithelial cancer cells and neural progenitor cells derived from patients with fragile X syndrome