Learn More
Research Products International Corp Cell Spreaders, L Shaped, Sterile, Individually Wrapped, 500 per Case
SDP
Catalog No. 50550278
Encompass
These cell spreaders have an L shape that provides quick and uniform cell distribution across surface of growth media. Spreaders are made from durable polystyrene for easy disposal after each use. Supplied in sealed pouches, sterile.
Catalog No. 50-550-278 Supplier Research Products International Corp Supplier No. 247662
These cell spreaders have an L shape that provides quick and uniform cell distribution across surface of growth media. Spreaders are made from durable polystyrene for easy disposal after each use. Supplied in sealed pouches, sterile.