Learn More
Selleck Chemical LLC LB-100 5mg 1026680-07-8
SDP
Catalog No. 501364915
Encompass_Preferred
LB-100 is a water soluble protein phosphatase 2A (PP2A) inhibitor with IC50s of 0.85 ?M and 3.87 ?M in BxPc-3 and Panc-1 cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-4915 Supplier Selleck Chemical LLC Supplier No. S75375MG
Add to Cart
Add to Cart
LB-100 is a water soluble protein phosphatase 2A (PP2A) inhibitor with IC50s of 0.85 ?M and 3.87 ?M in BxPc-3 and Panc-1 cells. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.