Learn More
Selleck Chemical LLC LB-100
SDP
Catalog No. 5000000068
Encompass_Preferred
LB-100 is a water soluble protein phosphatase 2A (PP2A) inhibitor with IC50s of 0 85 M and 3 87 M in BxPc-3 and Panc-1 cells
Catalog No. 50-000-00068 Supplier Selleck Chemical LLC Supplier No. S7537100MG
LB-100 is a water soluble protein phosphatase 2A (PP2A) inhibitor with IC50s of 0 85 M and 3 87 M in BxPc-3 and Panc-1 cells