Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Fischer Technical Company™ LB Agar, 200 mL
SCI_ED

Catalog No. S012389 Shop All Fischer Technical Company Products
Change view
Click to view available options
Quantity:
6 Bottles/Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S012389 6 Bottles/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Prop 65: Cancer and Reproductive Harm.
Catalog No. S012389 Supplier Fischer Technical Company™ Supplier No. CM945PB2006
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Prop 65: Cancer and Reproductive Harm.

Bottle of easy-to-pour and store LB Agar.

  • Each bottle is labeled with product name, expiration date and lot number for easy handling
  • Each plastic bottle contains 200 mL of media
  • Allows for easy pouring of plates
  • Sterile

Specifications

Detects Bacteria
Form Prepared Medium
Format Bottled
Manufacturing Origin US with Globally Sourced Materials
Media Type LB Agar
Product Type Microbiology Media
Sterility Sterile
Target Organism Class E. coli
Test Type Microbiological Tests
For Use With (Application) Microbiological Testing
Quantity 6 Bottles/Pk.
Volume (Metric) Specimen 200 mL
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.