Learn More
LAMDA BIOTECH INC LB Agar
Catalog No. 502045827
Encompass_Preferred
Premixed LB Agar: weigh it out, put it in the flask or bottle, add the appropriate volume of water and autoclave as usual.
Catalog No. 50-204-5827 Supplier LAMDA BIOTECH INC Supplier No. C121
Premixed LB Agar: weigh it out, put it in the flask or bottle, add the appropriate volume of water and autoclave as usual.