Learn More
Sigma Aldrich Fine Chemicals Biosciences Agar Ampicillin-100, Plates, 10pk
Catalog No. NC1032374
Encompass
Pre-poured agar plates with 100 ug/ml ampicillin. Miller’s LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids, water-soluble vitamins, and carbohydrates in a low-salt formulation. The addition of agar provides a solid medium for microbial growth.
Catalog No. NC1032374 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L566710EA
Pre-poured agar plates with 100 ug/ml ampicillin. Miller’s LB is a highly-referenced microbial growth medium used for the cultivation of E. coli. This nutrient-rich microbial broth contains peptides, amino acids, water-soluble vitamins, and carbohydrates in a low-salt formulation. The addition of agar provides a solid medium for microbial growth.