Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MP Biomedicals™ LB-Agar, Capsules, Molecular Biology Certified™
GSA_VA

Catalog No. MP113002221 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
11 g
227 g
454 g
Format:
Capsule
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Format
MP113002221 454 g Capsule
MP113002211 227 g Capsule
MP113002201 11 g Capsule
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP113002221 Supplier MP Biomedicals™ Supplier No. 113002221
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rich medium for the growth of E. coli and certified for plasmid and other bacterial cultures in liquid medium.

  • Nutritionally rich medium originally developed for growth and maintenance of E. coli strains
  • Eliminates the waste, inaccuracies, and mess associated with weighing out bulk powder
  • Contains essential electrolytes for transport and osmotic balance, due to NaCl component

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance Molecular Biology Certified™ FDA
Content And Storage 15°C to 30°C
Format Capsule
Grade Molecular Biology
Product Type Agar
Formulation Tryptone 10g/L, Yeast extract 5g/L, NaCl 10g/L, Agar 15g/L
For Use With (Application) For growth of bacteria
Quantity 454 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.