Learn More
Sigma Aldrich Fine Chemicals Biosciences LB Agar Carbenicillin - 100, Plates pre-poured agar plates with 100 mug/ml carbenicillin | 10EA
Catalog No. NC2095992
Encompass
LB Agar Carbenicillin - 100, Plates pre-poured agar plates with 100 mug/ml carbenicillin | 10EA
This item is not returnable. View return policy
Catalog No. NC2095992 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. L041810EA
LB Agar Carbenicillin - 100, Plates pre-poured agar plates with 100 mug/ml carbenicillin | 10EA