Learn More
Biotang Inc LB Agar, Lenox Agar, 1KG, P-820, CAS # Agar: 9002-18-0, CAS # Sodium Chloride: 7647-14-5, CAS # Yeast Extract: 8013-01-2. For preparation and maintenance of E. coli, 1.0 % Trypton, 0.5% Yeast Extract, 0.5% NaCl, and 1.5 % Agar.
Catalog No. 50152359 Shop All BIOTANG Inc Products
Encompass_Preferred
LB Agar, Lenox Agar, 1KG, P-820, CAS # Agar: 9002-18-0, CAS # Sodium Chloride: 7647-14-5, CAS # Yeast Extract: 8013-01-2. For preparation and maintenance of E. coli, 1.0 % Trypton, 0.5% Yeast Extract, 0.5% NaCl, and 1.5 % Agar.
Catalog No. 50-152-359 Supplier Biotang Inc Supplier No. BTBB905
LB Agar, Lenox Agar, 1KG, P-820, CAS # Agar: 9002-18-0, CAS # Sodium Chloride: 7647-14-5, CAS # Yeast Extract: 8013-01-2. For preparation and maintenance of E. coli, 1.0 % Trypton, 0.5% Yeast Extract, 0.5% NaCl, and 1.5 % Agar.