Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Research Products International Corp LB Agar, Low Salt Formula, Powder [Lennox L Agar], 500 Grams
SDP

Catalog No. 50213402
Encompass_Preferred

Molecular Biology Grade. For propagation and maintenance of E. Coli. Low Salt Formula.

Dissolve 32 grams in 1 Liter distilled water. Adjust the pH to 7.2 Sterilize by autoclaving for 15 minutes

Ingredients:

Casein Peptone: 10 g/L
Sodium Chloride: 5 g/L
Yeast Extract: 5 g/L
Agar: 12 g/L

Catalog No. 50-213-402 Supplier Research Products International Corp Supplier No. L24030500.0
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Molecular Biology Grade. For propagation and maintenance of E. Coli. Low Salt Formula.

Dissolve 32 grams in 1 Liter distilled water. Adjust the pH to 7.2 Sterilize by autoclaving for 15 minutes

Ingredients:

Casein Peptone: 10 g/L
Sodium Chloride: 5 g/L
Yeast Extract: 5 g/L
Agar: 12 g/L

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.