Learn More
Alkali Scientific LB Agar, Low Salt Formula, Powder [Lennox Agar], 2 Kg
SDP
Catalog No. 5000659744
Encompass_Preferred
LB Agar (Low Salt Formula) is designed to grow bacteria under conditions where reduced sodium is needed. The formulation comes in a powder form, making it easy to prepare by adding water. With a low salt concentration, this medium is ideal for culturing organisms that are sensitive to high levels of sodium. The 2kg size is convenient for medium to large-scale laboratories and industrial applications. The agar supports bacterial growth by providing nutrients such as dextrose, peptone, and vitamins. LB Agar is widely used in microbiology, molecular biology, and biotechnology, particularly in bacterial cloning, transformation, and expression studies.
Catalog No. 50-006-59744 Supplier Alkali Scientific Supplier No. LBA007CP
LB Agar (Low Salt Formula) is designed to grow bacteria under conditions where reduced sodium is needed. The formulation comes in a powder form, making it easy to prepare by adding water. With a low salt concentration, this medium is ideal for culturing organisms that are sensitive to high levels of sodium. The 2kg size is convenient for medium to large-scale laboratories and industrial applications. The agar supports bacterial growth by providing nutrients such as dextrose, peptone, and vitamins. LB Agar is widely used in microbiology, molecular biology, and biotechnology, particularly in bacterial cloning, transformation, and expression studies.