Learn More
Alkali Scientific LB Agar, Low Salt Formula, Powder [Lennox Agar], 500 G
SDP
Supplier:  Alkali Scientific LBA008CP
Encompass_Preferred
LB Agar (Low Salt Formula) Powder is a modified version of standard LB agar with reduced sodium content, making it suitable for culturing bacteria that are sensitive to high salt concentrations. The 500g powder can be easily dissolved in water to create a growth medium that supports bacterial culture by providing essential nutrients like dextrose, peptone, and vitamins. This medium is often used in molecular biology and biotechnology labs for bacterial cloning, transformation, and protein expression. The low-salt formula allows for optimal bacterial growth without the adverse effects of excess sodium, making it ideal for specific bacterial strains or sensitive applications.
Catalog No. 50-006-59963