Learn More
Teknova LB Agar Media, 1 Kg
SDP
Catalog No. 502388119
Encompass
1.0% Tryptone, 0.5% Yeast Extract,1.0% Sodium Chloride, 1.5% Agar
Catalog No. 50-238-8119 Supplier Teknova Supplier No. L9110
1.0% Tryptone, 0.5% Yeast Extract,1.0% Sodium Chloride, 1.5% Agar