Learn More
Teknova LB Agar Media. 454 grams
SDP
Catalog No. 50842668
Encompass
1.0% Tryptone, 0.5% Yeast Extract,1.0% Sodium Chloride, 1.5% Agar
Catalog No. 50-842-668 Supplier Teknova Supplier No. L9105
1.0% Tryptone, 0.5% Yeast Extract,1.0% Sodium Chloride, 1.5% Agar