Learn More
NEOGEN LB Agar, Miller, 5 kg
SDP
Catalog No. 502015562
Encompass_Preferred
LB Agar (Miller), a nutritionally rich medium designed for growth of pure cultures of recombinant strains, is based on the formulation described by Miller. E. coli is grown to late log phase in LB Medium. Some plasmid vectors replicate to a high copy number and do not require selective amplification. Some vectors do not replicate so freely and need to be selectively amplified. Chloramphenicol may be added to inhibit host synthesis and prevent replication of the bacterial chromosome.LB Agar, Miller contains 10 g/L of sodium chloride, different from the levels in Lennox formulations. This allows the researcher to select the optimal salt concentration for a specific strain. The medium may be aseptically supplemented with glucose or antibiotics.For more information visit https://www.neogen.com/solutions/Microbiology/lb-agar-miller
Catalog No. 50-201-5562 Supplier NEOGEN Supplier No. 700003377
LB Agar (Miller), a nutritionally rich medium designed for growth of pure cultures of recombinant strains, is based on the formulation described by Miller. E. coli is grown to late log phase in LB Medium. Some plasmid vectors replicate to a high copy number and do not require selective amplification. Some vectors do not replicate so freely and need to be selectively amplified. Chloramphenicol may be added to inhibit host synthesis and prevent replication of the bacterial chromosome.LB Agar, Miller contains 10 g/L of sodium chloride, different from the levels in Lennox formulations. This allows the researcher to select the optimal salt concentration for a specific strain. The medium may be aseptically supplemented with glucose or antibiotics.For more information visit https://www.neogen.com/solutions/Microbiology/lb-agar-miller