Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals LB Agar plate, Plain

Catalog No. aaj61525eqf
Change view
Click to view available options
Quantity:
20 Plates
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAJ61525EQF 20 Plates
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAJ61525EQF Supplier Thermo Scientific Chemicals Supplier No. J61525EQF
Only null left
Add to Cart

LB agar plate, also known as Luria Bertani medium with agar, is used to grow Escherichia coli and other bacterial cultures.

A subtype of nutrient agar, this is the general medium for microbiology studies and may be used for routine cultivation of not particularly fastidious microorganisms. Also, does not preferentially grow one kind of bacteria over another.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

• LB agar plate is a nutrient-rich media frequently used to culture bacteria in the lab, including Escherichia coli

Application

• LB agar plate is used to culture bacteria in microbiology research. These plates are commonly used during cloning experiments to amplify antibiotic-resistant, competent bacteria
• The addition of agar to LB medium helps form a gel that allows bacteria to grow. The bacteria cannot digest the agar but can obtain the needed nutrients to grow from the LB medium
• The addition of ampicillin (100 μg/mL) to these plates allows selection of the bacteria that can grow in the presence of ampicillin which is conferred by a plasmid carrying the antibiotic resistance gene

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage 2°C to 8°C
Form Solid
Media Type LB Agar
For Use With (Application) For growth of Escherichia coli, plasmid and other bacterial cultures
Packaging Type Plate
Quantity 20 Plates
Height (Metric) 15 mm
Diameter (Metric) Outside 10 cm

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.