Learn More
Teknova LB Luria Agar Plates with 1.6% Skim Milk. 100mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. NC0763967
Encompass
1.00% Tryptone, 0.50% Yeast Extract, 0.05% Sodium Chloride, 1.60% Skim Milk, 1.50% Agar
This item is not returnable. View return policy
Catalog No. NC0763967 Supplier Teknova Supplier No. L7614
1.00% Tryptone, 0.50% Yeast Extract, 0.05% Sodium Chloride, 1.60% Skim Milk, 1.50% Agar