Learn More
Teknova LB Agar Plates 1% Glucose and Kanamycin-50. 150mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502387536
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,1.0% Glucose, 50g/mL Kanamycin Sulfate
Catalog No. 50-238-7536 Supplier Teknova Supplier No. L5825
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,1.0% Glucose, 50g/mL Kanamycin Sulfate