Learn More
Teknova LB Agar Plates. 100mm Plates, 20 Plates per Sleeve, Sterile.
SDP
Supplier:  Teknova L1100
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar
Catalog No. 50-238-8202