Learn More
Teknova LB AGAR PLATES 100MM PLT 20PK
SDP
Supplier:  Teknova L1919
Encompass
LB Agar Plates with Gent-7, Kan-50, Tet-10 ,Bluo-gal-100 & IPTG-40. 100mm, 20 Plates per Sleeve
Catalog No. 50-842-017