Learn More
Teknova LB Agar Plates. 150mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502388106
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar
Catalog No. 50-238-8106 Supplier Teknova Supplier No. L5100
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar