Learn More
Kd Medical Inc LB AGAR PLATES
SDP
Catalog No. 501003475
Encompass_Preferred
LB Agar Plates. Used for the maintenance and propagation of E. coli. Ten 100mm plates per sleeve.
Catalog No. 50-100-3475 Supplier Kd Medical Inc Supplier No. BPL2010
LB Agar Plates. Used for the maintenance and propagation of E. coli. Ten 100mm plates per sleeve.