Learn More
Teknova LB Agar Plates with Ampicillin-100, X-Gal, and IPTG. 100mm, 20 Plates/Sleeve, Sterile. 8 Pack.
SDP
Catalog No. 502387555
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Ampicillin Sodium Salt, 60g/mL X-Gal,0.1mM Isopropyl-B-D-thiogalactopyranoside(IPTG)
Catalog No. 50-238-7555 Supplier Teknova Supplier No. L19028PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,100g/mL Ampicillin Sodium Salt, 60g/mL X-Gal,0.1mM Isopropyl-B-D-thiogalactopyranoside(IPTG)