Learn More
Teknova LB Agar Plates w/ Carbenicillin-50, X-Gal and IPTG. 100mm, 20 Plates per Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387456
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 50g/mL Carbenicillin Disodium Salt, 60g/mL X-Gal, 0.1mM Isopropyl-B-D-thiogalactopyranoside(IPTG)
Catalog No. 50-238-7456 Supplier Teknova Supplier No. L19048PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 50g/mL Carbenicillin Disodium Salt, 60g/mL X-Gal, 0.1mM Isopropyl-B-D-thiogalactopyranoside(IPTG)