Learn More
Teknova LB Agar Plates with Carbenicillin-50 and Kanamycin-25. 100mm, 20 plates/sleeve, Sterile. 8 Pack.
SDP
Catalog No. 502387812
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/mL Carbenicillin Disodium Salt,25g/mL Kanamycin Sulfate
Catalog No. 50-238-7812 Supplier Teknova Supplier No. L12188PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/mL Carbenicillin Disodium Salt,25g/mL Kanamycin Sulfate