Learn More
Teknova LB Agar Plates with Gentamicin-7, Kanamycin-50, Bluo-gal-100, IPTG-40. 100mm, 20 Plates/Sleeve, Sterile. 8 Pack.
SDP
Catalog No. 502387897
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 7g/mL Gentamycin, 50g/mL Kanamycin Sulfate, 100g/mL Bluo-gal, 40g/mL Isopropyl-B-D-thiogalactopyranoside(IPTG)
Catalog No. 50-238-7897 Supplier Teknova Supplier No. L19318PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 7g/mL Gentamycin, 50g/mL Kanamycin Sulfate, 100g/mL Bluo-gal, 40g/mL Isopropyl-B-D-thiogalactopyranoside(IPTG)