Learn More
Teknova LB Agar Plates with Gentamicin-7, Kan-50, Tet-10, X-Gal & IPTG. 100mm, 20 Plates/Sleeve, Sterile. 8 Pack.
SDP
Catalog No. 502387836
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/mL Kanamycin Sulfate, 7g/mL Gentamicin Sulfate, 10g/mL Tetracycline Hydrochloride,60g/mL X-Gal, 0.1mM Isopropyl-B-D-thiogalactopyranoside(IPTG)
Catalog No. 50-238-7836 Supplier Teknova Supplier No. L19188PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/mL Kanamycin Sulfate, 7g/mL Gentamicin Sulfate, 10g/mL Tetracycline Hydrochloride,60g/mL X-Gal, 0.1mM Isopropyl-B-D-thiogalactopyranoside(IPTG)