Learn More
Teknova LB Agar Plates with 1% Glucose and Kan-50. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387450
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,1.0% Cerelose Anhydrous Dextrose (Glucose), 50g/mL Kanamycin Sulfate
Catalog No. 50-238-7450 Supplier Teknova Supplier No. L77474PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,1.0% Cerelose Anhydrous Dextrose (Glucose), 50g/mL Kanamycin Sulfate