Learn More
Teknova LB Agar Plates with 1% Glucose and Kanamycin-30. 100mm, 20 Plates per Sleeve, Sterile.
SDP
Catalog No. 502387981
Encompass
1% Tryptone, 0.5% Yeast extract, 1.0% Sodium chloride, 1.5% Agar,1.0% Glucose, 30g/mL Kanamycin Sulfate
Catalog No. 50-238-7981 Supplier Teknova Supplier No. L1830
1% Tryptone, 0.5% Yeast extract, 1.0% Sodium chloride, 1.5% Agar,1.0% Glucose, 30g/mL Kanamycin Sulfate