Learn More
Teknova LB Agar Plates with 1% Glucose and Chloramphenicol-34. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387348
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium chloride, 1.5% Agar,1.0% Cerelose Anhydrous Dextrose (Glucose), 34g/mL Chloramphenicol
Catalog No. 50-238-7348 Supplier Teknova Supplier No. L18074PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium chloride, 1.5% Agar,1.0% Cerelose Anhydrous Dextrose (Glucose), 34g/mL Chloramphenicol