Learn More
Teknova LB Agar Plates with 2% Glucose and Carbenicillin-100. 150mm, 20 Plates/Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387522
Encompass
1.0% Tryptone, 0.5% Yeast extract, 1.0% Sodium chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt, 2% Cerelose Anhydrous Dextrose (Glucose)
Catalog No. 50-238-7522 Supplier Teknova Supplier No. L58028PK
1.0% Tryptone, 0.5% Yeast extract, 1.0% Sodium chloride, 1.5% Agar,100g/mL Carbenicillin Disodium Salt, 2% Cerelose Anhydrous Dextrose (Glucose)