Learn More
Teknova LB Agar Plates with 2% Glucose and Carbenicillin-100, 245mm, 4 Plates/Sleeve, Sterile. 8 Pack.
SDP
Catalog No. 502387912
Encompass
1.0% Tryptone, 0.5% Yeast extract, 1.0% Sodium chloride, 2.0% Cerelose Anhydrous Dextrose (Glucose), 1.5% Agar, 100g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-7912 Supplier Teknova Supplier No. L68988PK
1.0% Tryptone, 0.5% Yeast extract, 1.0% Sodium chloride, 2.0% Cerelose Anhydrous Dextrose (Glucose), 1.5% Agar, 100g/mL Carbenicillin Disodium Salt