Learn More
Teknova LB Agar Plates with 5% Sucrose and Carb-100. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387810
Encompass
1.0% Tryptone, 0.5% Yeast extract, 1.0% Sodium Chloride, 1.5% Agar,5.0% Sucrose,100g/mL Carbenicillin Disodium Salt
Catalog No. 50-238-7810 Supplier Teknova Supplier No. L18654PK
1.0% Tryptone, 0.5% Yeast extract, 1.0% Sodium Chloride, 1.5% Agar,5.0% Sucrose,100g/mL Carbenicillin Disodium Salt