Learn More
Teknova LB Agar Plates with 6% Sucrose without Sodium Chloride. 100mm, 20 Plates per Sleeve, Sterile. 4 pack.
SDP
Catalog No. 502387266
Encompass
1.0% Tryptone, 0.5% Yeast Extract,1.5% Agar, 6.0% Sucrose
Catalog No. 50-238-7266 Supplier Teknova Supplier No. L11434PK
1.0% Tryptone, 0.5% Yeast Extract,1.5% Agar, 6.0% Sucrose