Learn More
Teknova LB Agar Plates with Animal-Free Soytone, Chloramphenicol-34, Kanamycin-50. 100mm, 20 Plates per Sleeve, Sterile. 4 Pack.
SDP
Catalog No. 502387643
Encompass
1.69% Animal-Free Soytone, 0.50% Yeast Extract, 1.00% Sodium Chloride, 1.50% Agar, 34g/mL Chloramphenicol, 50g/mL Kanamycin Sulfate
Catalog No. 50-238-7643 Supplier Teknova Supplier No. L15724PK
1.69% Animal-Free Soytone, 0.50% Yeast Extract, 1.00% Sodium Chloride, 1.50% Agar, 34g/mL Chloramphenicol, 50g/mL Kanamycin Sulfate