Learn More
Teknova LB Agar Plates with Amp-100. Vented QTrays. 245X245mm, 4 Plates per Sleeve, Sterile. 8 pack.
SDP
Catalog No. 502387504
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 100g/mL Ampicillin Sodium Salt
Catalog No. 50-238-7504 Supplier Teknova Supplier No. L40048PK
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar, 100g/mL Ampicillin Sodium Salt