Learn More
Teknova LB Agar Plates with Ampicillin-50. 100mm, 10 plates per Sleeve, Sterile. 8 Pack.
SDP
Supplier:  Teknova L11508PK
Encompass
1.0% Tryptone, 0.5% Yeast Extract, 1.0% Sodium Chloride, 1.5% Agar,50g/mL Ampicillin Sodium Salt
Catalog No. 50-238-7632